Novus Biologicals
Manufacturer Code:NBP2822790.1ML
Catalog # NB030208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human MAFG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic leucine zipper transcription factor MafG; basic leucine zipper transcription factor MafG hMAF MGC13090 MGC20149 transcription factor MafG v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family protein G V-maf musculoaponeurotic fibrosarcoma oncogene homolog G v-maf musculoaponeurotic fibrosarcoma oncogene homolog G (avian); hMAF; Transcription factor MafG; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog G; V-maf musculoaponeurotic fibrosarcoma oncogene homolog G
Gene Aliases: hMAF; MAFG
UniProt ID: (Human) O15525
Entrez Gene ID: (Human) 4097
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.