Invitrogen
Manufacturer Code:PA524019
Catalog # PIPA524019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 111-139 amino acids from the N-terminal region of human MYSM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2A-DUB; 2A-DUB 2ADUB RP4-592A1.1 histone H2A deubiquitinase MYSM1 myb-like SWIRM and MPN domain-containing protein 1; Deubiquitinase MYSM1; histone H2A deubiquitinase MYSM1; Myb-like, SWIRM and MPN domain-containing protein 1; Myb-like, SWIRM and MPN domains 1
Gene Aliases: 2A-DUB; 2ADUB; C130067A03Rik; C530050H10Rik; KIAA1915; MYSM1
UniProt ID: (Human) Q5VVJ2, (Mouse) Q69Z66
Entrez Gene ID: (Human) 114803, (Mouse) 320713
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.