Proteintech
Manufacturer Code:200781AP150UL
Catalog # 50-173-2686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2A DUB 2ADUB KIAA1915 MYSM1 RP4 592A1.1; 2A-DUB; Deubiquitinase MYSM1; Myb-like, SWIRM and MPN domain-containing protein 1; Myb-like, SWIRM and MPN domains 1
Gene Aliases: 2A-DUB; 2ADUB; KIAA1915; MYSM1
UniProt ID: (Human) Q5VVJ2
Entrez Gene ID: (Human) 114803
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.