Novus Biologicals
Manufacturer Code:NBP287870
Catalog # NB035492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human MYRIP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp586F1018 exophilin-8 FLJ44025 MGC130034 myosin VIIA and Rab interacting protein Myosin-VIIa- and Rab-interacting protein MyRIP rab effector MyRIP SLAC2-C SLAC2CMGC130035 Slp homolog lacking C2 domains c Slp homologue lacking C2 domains synaptotagmin-like protein homologue lacking C2 domains-c; Exophilin-8; Myosin-VIIa- and Rab-interacting protein; Rab effector MyRIP; SlaC2-c; slp homolog lacking C2 domains c; Slp homologue lacking C2 domains; synaptotagmin-like protein homologue lacking C2 domains-c; Synaptotagmin-like protein lacking C2 domains C
Gene Aliases: MYRIP; SLAC2-C; SLAC2C
UniProt ID: (Human) B7Z9V3
Entrez Gene ID: (Human) 25924
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cytoskeletal protein enzyme modulator non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.