Invitrogen
Manufacturer Code:PA517164
Catalog # PIPA517164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide correspondent to amino-terminal residues of human MYPT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 130 kDa myosin-binding subunit of smooth muscle myosin phophatase; 130 kDa myosin-binding subunit of smooth muscle myosin phosphatase; 130 kDa regulatory subunit of myosin phosphatase; 133kDa myosin-binding subunit of smooth muscle myosin phosphatase (M133); MBSP; myosin binding subunit; Myosin phosphatase target subunit 1; myosin phosphatase target subunit 1 myosin phosphatase-targeting subunit 1 protein phosphatase 1 regulatory subunit 12A protein phosphatase myosin-binding subunit; myosin phosphatase, target subunit 1; Myosin phosphatase-targeting subunit 1; myosin-binding subunit of myosin phosphatase; mypt1; PP-1M; PP1M subunit M110; Protein phosphatase 1 regulatory subunit 12A; protein phosphatase 1, regulatory (inhibitor) subunit 12A; protein phosphatase 1, regulatory subunit 12A; protein phosphatase 1M 110 kda regulatory subunit; Protein phosphatase myosin-binding subunit; Protein phosphatase subunit 1M; Serine/threonine protein phosphatase PP1 smooth muscle regulatory subunit M110; smooth muscle myosin phosphatase myosin-binding subunit; unm p82emcf
Gene Aliases: 1200015F06Rik; 5730577I22Rik; AA792106; AV099298; D10Ertd625e; I79_016312; M110; M130; MBS; MBSP; MYPT1; PPP1R12A; si:dkey-28j4.1; unm_p82emcf; zgc:110448
UniProt ID: (Chicken) Q90623, (Human) O14974, (Rat) Q10728, (Mouse) Q9DBR7, (Zebrafish) Q6DRG7
Entrez Gene ID: (Pig) 397190, (Chicken) 396020, (Bovine) 538606, (Dog) 475411, (Human) 4659, (Rat) 116670, (Mouse) 17931, (Chinese hamster) 100754718, (Zebrafish) 445393
Molecular Function: enzyme modulator phosphatase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.