Invitrogen
Manufacturer Code:PA570479
Catalog # PIPA570479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human MYPN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 145 kDa sarcomeric protein; Myopalladin; sarcomeric protein myopalladin, 145 kDa (MYOP)
Gene Aliases: 1110056A04Rik; 9830112J03; AI853556; CMD1DD; CMH22; Kiaa4170; mKIAA4170; MYOP; MYPN; RCM4; wu:fc17a02
UniProt ID: (Human) Q86TC9, (Mouse) Q5DTJ9
Entrez Gene ID: (Human) 84665, (Pig) 100624901, (Rabbit) 100348592, (Dog) 489013, (Horse) 100072581, (Rat) 309760, (Mouse) 68802, (Guinea pig) 100734734, (Zebrafish) 571695
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.