OriGene
Manufacturer Code:TA808910
Catalog # 50-168-2461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C2 |
Immunogen | Human recombit protein fragment corresponding to amino acids 106-299 of human MYOZ1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Calsarcin-2; CS-2 FATZ MYOZ; Filamin-, actinin- and telethonin-binding protein; MYOZ1; Myozenin-1; Protein FATZ
Gene Aliases: CS-2; FATZ; MYOZ; MYOZ1
UniProt ID: (Human) Q9NP98
Entrez Gene ID: (Human) 58529
Molecular Function: structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.