Invitrogen
Manufacturer Code:PA569432
Catalog # PIPA569432
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human MYOF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: fer-1-like 3, myoferlin; fer-1-like family member 3; Fer-1-like protein 3; Myoferlin
Gene Aliases: 2310004N10Rik; 2310051D19Rik; E030042N20Rik; FER1L3; KIAA1207; MYOF; RGD1564216; zgc:63504
UniProt ID: (Human) Q9NZM1, (Mouse) Q8C0D1
Entrez Gene ID: (Rabbit) 100342909, (Human) 26509, (Bovine) 513597, (Horse) 100071199, (Dog) 607472, (Mouse) 226101, (Rat) 309499, (Zebrafish) 393849
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.