Proteintech
Manufacturer Code:189431AP150UL
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYOD1 Fusion Protein Ag13512 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHc1; bHLHc1 Myf 3 MYF3 MYOD MYOD1 myogenic differentiation 1 Myogenic factor 3 PUM; Class C basic helix-loop-helix protein 1; Myf-3; Myoblast determination protein 1; Myogenic factor 3
Gene Aliases: AI503393; BHLHC1; MYF3; MYOD; Myod-1; MYOD1; PUM
UniProt ID: (Human) P15172, (Mouse) P10085, (Rat) Q02346
Entrez Gene ID: (Human) 4654, (Mouse) 17927, (Rat) 337868
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.