Invitrogen
Manufacturer Code:MA512902
Catalog # PIMA512902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5.8A |
Immunogen | Recombit mouse MyoD1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHc1; Class C basic helix-loop-helix protein 1; class C basic helix-loop-helix protein 1 myf-3 myoblast determination protein 1 myogenic factor 3; Myf-3; Myoblast determination protein 1; Myoblast determination protein 1 homolog; MYOD1 homolog; myogenic factor 1; Myogenic factor 3; unnamed protein product
Gene Aliases: AI503393; BHLHC1; CMD1; MYF3; MYOD; Myod-1; MYOD1; PUM
UniProt ID: (Human) P15172, (Chicken) P16075, (Mouse) P10085, (Rat) Q02346
Entrez Gene ID: (Human) 4654, (Chicken) 374048, (Mouse) 17927, (Rat) 337868
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.