Invitrogen
Manufacturer Code:PA523078
Catalog # PIPA523078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an N-terminal region of the human MyoD1 protein (within residues 1-150). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bHLHc1; Class C basic helix-loop-helix protein 1; Myf-3; MYF3 MYOD PUM bHLHc1 class C basic helix-loop-helix protein 1 myf-3 myoblast determination protein 1 myogenic factor 3; Myoblast determination protein 1; Myogenic factor 3
Gene Aliases: BHLHC1; MYF3; MYOD; MYOD1; PUM
UniProt ID: (Human) P15172
Entrez Gene ID: (Human) 4654
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.