Novus Biologicals
Manufacturer Code:NB10041092
Catalog # NB10041092
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KNDFEPVKQSKAQ corresponding to internal region according to NP_004136.2 NP_001123537.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CELIAC4 myosin IXB myosin-IXb MYR5Unconventional myosin-9b unconventional myosin IXb; myosin-IXb; unconventional myosin IXb; Unconventional myosin-9b; Unconventional myosin-IXb
Gene Aliases: CELIAC4; MYO9B; MYR5
UniProt ID: (Human) Q13459
Entrez Gene ID: (Human) 4650
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.