Novus Biologicals
Manufacturer Code:NBP298029AF594
Catalog # NB051272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MYO9B fragment. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CELIAC4 myosin IXB myosin-IXb MYR5Unconventional myosin-9b unconventional myosin IXb; myosin-IXb; unconventional myosin IXb; Unconventional myosin-9b; Unconventional myosin-IXb
Gene Aliases: CELIAC4; MYO9B; MYR5
UniProt ID: (Human) Q13459
Entrez Gene ID: (Human) 4650
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.