Invitrogen
Manufacturer Code:PA576417
Catalog # PIPA576417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human MYO5B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1119 Unconventional myosin-Vb; MYO5B variant protein; myosin 5B; Myosin heavy chain myr 6; Myosin of the dilute-myosin-V family; myosin-Vb; Unconventional myosin-Vb
Gene Aliases: AI661750; KIAA1119; mKIAA1119; MYO5B; Myr6
UniProt ID: (Human) Q9ULV0, (Rat) P70569
Entrez Gene ID: (Human) 4645, (Mouse) 17919, (Rat) 25132
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.