Invitrogen
Manufacturer Code:PA532232
Catalog # PIPA532232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 1028 of Human Myosin 1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MMI-beta; MMI-beta MMIb NMI myr2 myosin I beta myosin-I beta myosin-Ic nuclear myosin I; Myosin I beta; myosin-I beta; myosin-Ic; nuclear myosin I; Unconventional myosin-Ic
Gene Aliases: MMI-beta; MMIb; MYO1C; myr2; NMI
UniProt ID: (Human) O00159
Entrez Gene ID: (Human) 4641
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.