Invitrogen
Manufacturer Code:PA529084
Catalog # PIPA529084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 299 and 547 of Human myosin IA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BBM-I; BBMI DFNA48 MIHC MYHL brush border myosin I myosin I heavy chain myosin heavy polypeptide-like (100kD) myosin-Ia; Brush border myosin I; MIHC; Myosin I heavy chain; myosin, heavy polypeptide-like (100kD); Unconventional myosin-Ia
Gene Aliases: BBMI; DFNA48; MIHC; MYHL; MYO1A
UniProt ID: (Human) Q9UBC5
Entrez Gene ID: (Human) 4640
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.