Proteintech
Manufacturer Code:188281AP150UL
Catalog # 50-173-2246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0865 MYO16 Myo16b myosin XVI MYR8 Unconventional myosin 16; myo16; MYO16 variant protein; myosin heavy chain Myr 8; myosin-XVI; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 3; Neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 3; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 3; protein phosphatase 1, regulatory subunit 107; Unconventional myosin-16; Unconventional myosin-XVI
Gene Aliases: BM140241; C230040D10Rik; Gm1105; KIAA0865; mKIAA0865; MYAP3; MYO16; MYO16B; MYR8; NYAP3; PPP1R107
UniProt ID: (Human) Q9Y6X6, (Mouse) Q5DU14
Entrez Gene ID: (Human) 23026, (Mouse) 244281
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.