Abnova
Manufacturer Code:PAB23644
Catalog # 89-125-353
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MYLK3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MYLK3.
For Research Use Only
Protein Aliases: caMLCK; Cardiac-MLCK; cardiac-MyBP-C associated Ca/CaM kinase; Cardiac-MyBP-C-associated Ca/CaM kinase; MGC126319; MGC126320; MLC kinase; MLCK; MLCK2; Myosin light chain kinase 3; putative myosin light chain kinase 3
Gene Aliases: caMLCK; MLCK; MLCK2; MYLK3
UniProt ID: (Human) Q32MK0
Entrez Gene ID: (Human) 91807
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.