Invitrogen
Manufacturer Code:PA589733
Catalog # PIPA589733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-90 of human MYLK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MLCK2; myosin light chain kinase 2, skeletal muscle; Myosin light chain kinase 2, skeletal/cardiac muscle
Gene Aliases: 9830004H17Rik; BB138278; KMLC; MLCK; MLCK2; MYLK2; skMLCK
UniProt ID: (Human) Q9H1R3, (Mouse) Q8VCR8
Entrez Gene ID: (Human) 85366, (Mouse) 228785
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.