Invitrogen
Manufacturer Code:PA529324
Catalog # PIPA529324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 270 and 551 of Human MYLK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KMLC MLCK MLCK2 skMLCK myosin light chain kinase 2 skeletal muscle myosin light chain kinase 2 skeletal/cardiac muscle; MLCK2; myosin light chain kinase 2, skeletal muscle; Myosin light chain kinase 2, skeletal/cardiac muscle
Gene Aliases: KMLC; MLCK; MLCK2; MYLK2; skMLCK
UniProt ID: (Human) Q9H1R3
Entrez Gene ID: (Human) 85366
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.