Novus Biologicals
Manufacturer Code:NBP211574
Catalog # NBP211574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KDTKNMEAKKLSKD corresponding to internal region according to NP_444253.3 NP_444254.3 NP_444255.3 NP_444256.3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.11 EC 2.7.11.18 KMLC MLCK2MLCK myosin light chain kinase 2 myosin light chain kinase 2 skeletal/cardiac muscle skeletal muscle skMLCK; MLCK2; myosin light chain kinase 2, skeletal muscle; Myosin light chain kinase 2, skeletal/cardiac muscle
Gene Aliases: KMLC; MLCK; MLCK2; MYLK2; skMLCK
UniProt ID: (Human) Q9H1R3
Entrez Gene ID: (Human) 85366
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.