Abnova
Manufacturer Code:MAB19490
Catalog # 89-336-015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | G-13 |
Immunogen | A synthetic peptide corresponding to human MYLK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C for one year. After reconstitution at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
Rabbit Monoclonal Antibody.
Protein Aliases: DKFZp686I10125 FLJ12216 KRP MLCK MLCK1 MLCK108 MLCK210 MSTP083 MYLK1 smMLCK; Kinase-related protein; KRP; MLCK; Myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; smooth muscle myosin light chain kinase; Telokin
Gene Aliases: AAT7; KRP; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; MYLK; MYLK1; smMLCK
UniProt ID: (Human) Q15746
Entrez Gene ID: (Human) 4638
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.