Proteintech
Manufacturer Code:216421AP150UL
Catalog # 50-173-3530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYLK Fusion Protein Ag16306 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kinase related protein KRP MLCK MLCK1 MLCK108 MLCK210 MSTP083 MYLK MYLK1 myosin light chain kinase smMLCK Telokin; Kinase-related protein; KRP; MLCK; mylk; Myosin light chain kinase, smooth muscle; Myosin light chain kinase, smooth muscle, deglutamylated form; myosin, light polypeptide kinase; smooth muscle myosin light chain kinase; Telokin
Gene Aliases: 9530072E15Rik; A930019C19Rik; AAT7; AW489456; KRP; MLCK; MLCK1; MLCK108; MLCK210; MSTP083; MYLK; MYLK1; nmMlck; smMLCK
UniProt ID: (Human) Q15746
Entrez Gene ID: (Human) 4638, (Mouse) 107589, (Rat) 288057
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.