Novus Biologicals
Manufacturer Code:NBP154903
Catalog # NBP154903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MYLIP(myosin regulatory light chain interacting protein) The peptide sequence was selected from the middle region of MYLIP. Peptide sequence CSSCEGLSCQQTRVLQEKLRKLKEAMLCMVCCEEEINSTFCPCGHTVCCE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: band 4.1 superfamily member BZF1; band 4.1 superfamily member BZF1 BZF1 cellular modulator of immune recognition (c-MIR) EC 6.3.2.- Idol IDOLmyosin regulatory light chain-interacting protein Inducible degrader of the LDL-receptor MIRE3 ubiquitin-protein ligase MYLIP myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; Idol; Inducible degrader of the LDL-receptor; MIR; Myosin regulatory light chain interacting protein; myosin regulatory light chain-interacting protein; RING-type E3 ubiquitin transferase MYLIP
Gene Aliases: BM-023; BZF1; IDOL; MIR; MYLIP; PP5242
UniProt ID: (Human) Q8WY64
Entrez Gene ID: (Human) 29116
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.