Proteintech
Manufacturer Code:154551AP150UL
Catalog # 50-173-0241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYLIP Fusion Protein Ag7720 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: band 4.1 superfamily member BZF1; BZF1 Idol MIR MYLIP PP5242; cellular modulator of immune recognition (c-MIR); E3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor; E3 ubiquitin-protein ligase MYLIP; Idol; Inducible degrader of the LDL-receptor; MIR; Myosin regulatory light chain interacting protein; Myosin regulatory light chain-interacting protein; RING-type E3 ubiquitin transferase MYLIP
Gene Aliases: 9430057C20Rik; BM-023; BZF1; I79_001311; IDOL; MIR; MYLIP; PP5242
UniProt ID: (Human) Q8WY64, (Mouse) Q8BM54, (Rat) D3ZDI6
Entrez Gene ID: (Human) 29116, (Chinese hamster) 100760940, (Mouse) 218203, (Rat) 306825
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.