Invitrogen
Manufacturer Code:PA517624
Catalog # PIPA517624
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino-terminal residues of human Myosin Light Chain 2 (smooth muscle isoform) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20 kDa myosin light chain; 20 kDa myosin light chain MLC-2C myosin RLC myosin regulatory light chain 1 myosin regulatory light chain 2 smooth muscle isoform myosin regulatory light chain 9 myosin regulatory light chain MRLC1 myosin regulatory light polypeptide 9 myosin light polypeptide 9 regulatory; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MLC-2; MLC-2C; myosin light chain 2; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; smooth muscle major isoform; unnamed protein product
Gene Aliases: AI327049; DTNB; G1; LC20; MLC-2; MLC-2C; MLC2; MLC20; MRLC1; MYL9; Mylc2c; MYRL2; RLC-C
UniProt ID: (Pig) P29269, (Chicken) P02612, (Human) P24844, (Rat) Q64122, (Mouse) Q3TW15
Entrez Gene ID: (Pig) 100157760, (Chicken) 396215, (Bovine) 505964, (Human) 10398, (Rat) 296313, (Mouse) 98932
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.