Novus Biologicals
Manufacturer Code:NBP15538320UL
Catalog # NBP15538320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MYL9(myosin light chain 9 regulatory) The peptide sequence was selected from the middle region of MYL9. Peptide sequence FDEEASGFIHEDHLRELLTTMGDRFTDEEVDEMYREAPIDKKGNFNYVEF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20 kDa myosin light chain; LC20; LC2020 kDa myosin light chain MLC2Myosin RLC MRLC1MLC-2C myosin regulatory light chain 1 Myosin regulatory light chain 2 smooth muscle isoform Myosin regulatory light chain 9 Myosin regulatory light chain MRLC1 myosin regulatory light polypeptide 9 myosin light chain 9 regulatory myosin light polypeptide 9 regulatory MYRL2MGC3505; MLC-2C; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory
Gene Aliases: LC20; MLC-2C; MLC2; MRLC1; MYL9; MYRL2
UniProt ID: (Human) P24844
Entrez Gene ID: (Human) 10398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.