Proteintech
Manufacturer Code:153541AP150UL
Catalog # 50-173-0171
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYL9 Fusion Protein Ag7600 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 20 kDa myosin light chain; 20 kDa myosin light chain LC20 MLC 2C MLC2 MRLC1 MYL9 Myosin RLC MYRL2; LC20; MLC-2C; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light polypeptide 9; Myosin RLC; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory
Gene Aliases: AI327049; LC20; MLC-2C; MLC2; MLC20; MRLC1; MYL9; Mylc2c; MYRL2; RLC-C
UniProt ID: (Human) P24844, (Rat) Q64122, (Mouse) Q3TW15
Entrez Gene ID: (Human) 10398, (Rat) 296313, (Mouse) 98932
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.