Proteintech
Manufacturer Code:109131AP150UL
Catalog # 50-172-6707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYL3 Fusion Protein Ag1364 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac myosin light chain 1; Cardiac myosin light chain 1 CMH8 CMLC1 MLC1SB MLC1V MYL3 Myosin light chain 3 VLC1; CMLC1; MLC-lV/sb; MLC1SB; MLClV; Myosin alkali light chain 1, ventricular; Myosin alkali light chain 1, ventricular/slow skeletal muscle isoform; Myosin light chain 1, slow-twitch muscle B/ventricular isoform; Myosin light chain 3; Myosin light chain 3, alkali, cardiac ventricles; myosin light chain, alkali, cardiac ventricles; myosin, light chain 3, alkali; ventricular, skeletal, slow; myosin, light polypeptide 3; myosin, light polypeptide 3, alkali; ventricular, skeletal, slow; rVMLC1; slow skeletal; ventricular; Ventricular myosin alkali light chain; Ventricular myosin light chain 1; ventricular, skeletal, slow; Ventricular/slow twitch myosin alkali light chain; VLCl
Gene Aliases: CMH8; MLC-lV/sb; MLC1s; MLC1SB; MLC1V; MYL3; Mylc; Mylc1v; VLC1; VLCl
UniProt ID: (Human) P08590, (Mouse) P09542, (Rat) P16409
Entrez Gene ID: (Human) 4634, (Mouse) 17897, (Rat) 24585
Molecular Function:
actin family cytoskeletal protein
calcium-binding protein
calmodulin
cytoskeletal protein
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.