Invitrogen
Manufacturer Code:PA5102917
Catalog # PIPA5102917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MYL12A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900073G15Rik AI838292 brain specific myosin regulatory light chain C80200 Epididymis secretory protein Li 24 HEL-S-24 MLC-2 MLC20 MLC-2A MLC-2B MLCB MRCL3 MRLC2 MRLC3 MYL12A MYL12B MYL2B MYLC2B myosin light chain 12A myosin light chain regulatory B-like myosin regulatory light chain 12A Myosin regulatory light chain 12B Myosin regulatory light chain 2 nonsarcomeric Myosin regulatory light chain 20 kDa myosin regulatory light chain 2-A smooth muscle isoform Myosin regulatory light chain 2-B smooth muscle isoform myosin regulatory light chain 3 Myosin regulatory light chain MRLC2 Myosin regulatory light chain MRLC3 myosin regulatory light chain RLC-A Myosin RLC myosin RLC-A myosin light chain 12A regulatory non-sarcomeric myosin light polypeptide regulatory non-sarcomeric (20kD) NMDA receptor-interacting protein RGD1309537 RLC Rlc-a SHUJUN-1; Epididymis secretory protein Li 24; HEL-S-24; MLC-2; MLC-2A; MLC-2B; MLC20; Myosin regulatory light chain 12A; Myosin regulatory light chain 12B; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2-B, smooth muscle isoform; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 3; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; Myosin RLC; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); SHUJUN-1
Gene Aliases: HEL-S-24; MLC-2B; MLCB; MRCL3; MRLC2; MRLC3; MYL12A; MYL12B; MYL2B; MYLC2B; RLC
UniProt ID: (Human) P19105
Entrez Gene ID: (Human) 10627
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.