Invitrogen
Manufacturer Code:PA571967
Catalog # PIPA571967
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 134-165 amino acids from the N-terminal region of human MYH9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BDPLT6 DFNA17 EPSTS FTNS MHA NMHC-II-A NMMHC-IIA NMMHCA cellular myosin heavy chain type A myosin heavy chain 9 myosin heavy chain non-muscle IIa myosin-9 non-muscle myosin heavy chain A non-muscle myosin heavy chain IIa non-muscle myosin heavy polypeptide 9 nonmuscle myosin heavy chain II-A; Cellular myosin heavy chain, type A; flectin; Myosin heavy chain 9; myosin heavy chain IX; Myosin heavy chain, non-muscle IIa; myosin IIA; Myosin-9; NMHC II-A; NMMHC II-a; NMMHC-A; non-muscle myosin heavy chain 9; Non-muscle myosin heavy chain A; Non-muscle myosin heavy chain IIa; non-muscle myosin heavy polypeptide 9; nonmuscle myosin heavy chain II-A
Gene Aliases: BDPLT6; DFNA17; EPSTS; Fltn; FTNS; MHA; MYH9; Myhn-1; Myhn1; NMHC-II-A; NMHCIIA; NMMHC-A; NMMHC-IIA; NMMHCA; TU72.6
UniProt ID: (Human) P35579, (Mouse) Q8VDD5
Entrez Gene ID: (Human) 4627, (Mouse) 17886
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.