Abnova
Manufacturer Code:PAB28470
Catalog # 89-129-977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MYH9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MYH9
For Research Use Only
Protein Aliases: Cellular myosin heavy chain, type A; DFNA17; EPSTS; FTNS; MGC104539; MHA; Myosin heavy chain 9; Myosin heavy chain, non-muscle IIa; Myosin-9; NMHC-II-A; NMMHC II-a; NMMHC-A; NMMHCA; non-muscle myosin heavy chain 9; Non-muscle myosin heavy chain A; Non-muscle myosin heavy chain IIa; non-muscle myosin heavy polypeptide 9; nonmuscle myosin heavy chain II-A
Gene Aliases: BDPLT6; DFNA17; EPSTS; FTNS; MHA; MYH9; NMHC-II-A; NMMHC-IIA; NMMHCA
UniProt ID: (Human) P35579
Entrez Gene ID: (Human) 4627
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.