Invitrogen
Manufacturer Code:PA569132
Catalog # PIPA569132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse MYH7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta myosin heavy chain; beta-myosin; beta-myosin heavy chain; beta-myosin-heavy-chain; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain beta isoform; CMH1 MPD1 SPMD SPMM CMD1S MYHCB; MYH-beta/slow; MyHC-beta; MyHC-slow; myopathy, distal 1; myosin beta heavy chain; Myosin heavy chain 7; myosin heavy chain cardiac alpha; myosin heavy chain polypeptide 7 cardiac muscle fetal; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 7; myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy polypeptide 7, cardiac muscle, beta; Myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A; type slow/beta myosin heavy chain; ventricular myosin heavy chain
Gene Aliases: B-MHC; beta-MHC; CMD1S; CMH1; mg:cb02g011; MPD1; MYH7; MyHC-alpha; Myhc-b; MyHC-I; MYHCB; smyhc3; SPMD; SPMM; vmhc
UniProt ID: (Horse) Q8MJU9, (Bovine) Q9BE39, (Human) P12883, (Dog) P49824, (Mouse) Q91Z83, (Rat) P02564
Entrez Gene ID: (Guinea pig) 100727138, (Rabbit) 100101566, (Horse) 791234, (Bovine) 282714, (Human) 4625, (Dog) 403807, (Mouse) 140781, (Rat) 29557, (Zebrafish) 30616
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.