Invitrogen
Manufacturer Code:PA5112257
Catalog # PIPA5112257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived Human MYH7 fragment |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta cardiac myosin heavy chain beta myosin heavy chain beta-MHC beta-myosin-heavy-chain B-MHC BOS_10147 cardiac muscle myosin heavy chain 7 beta cardiac myosin heavy chain beta isoform CMD1S CMH1 LOC100727138 mg:cb02g011 MPD1 MYH7 MYH-beta/slow MYHCB Myhc-b MyHC-beta MyHC-I MyHC-slow myopathy distal 1 myosin 7 myosin beta heavy chain Myosin heavy chain 7 myosin heavy chain polypeptide 7 cardiac muscle fetal Myosin heavy chain slow isoform myosin heavy chain slow type 1 (beta cardiac) Myosin heavy chain cardiac muscle beta isoform myosin heavy chain cardiac muscle fetal myosin heavy chain polypeptide 7 myosin heavy chain 7 cardiac muscle beta myosin heavy polypeptide 7 cardiac muscle beta Myosin-7 rhabdomyosarcoma antigen MU-RMS-40.7A smyhc3 SPMD SPMM type slow/beta myosin heavy chain ventricular myosin heavy chain vmhc; cardiac muscle myosin heavy chain 7 beta; MyHC-beta; MyHC-slow; myopathy, distal 1; Myosin heavy chain 7; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle beta isoform; myosin, heavy polypeptide 7, cardiac muscle, beta; Myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A
Gene Aliases: CMD1S; CMH1; MPD1; MYH7; MYHCB; SPMD; SPMM
UniProt ID: (Human) P12883
Entrez Gene ID: (Human) 4625
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.