Novus Biologicals
Manufacturer Code:NBP297775B
Catalog # NB103411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MYH7 fragment. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cardiac muscle beta isoform CMD1SMGC138376 CMH1 DKFZp451F047 MGC138378 MPD1 MYHCB MyHC-beta myhc-slow myopathy distal 1 myosin heavy chain (AA 1-96) myosin heavy chain 7 cardiac muscle beta myosin heavy polypeptide 7 cardiac muscle beta myosin-7 rhabdomyosarcoma antigen MU-RMS-40.7A SPMD SPMM; cardiac muscle myosin heavy chain 7 beta; MyHC-beta; MyHC-slow; myopathy, distal 1; Myosin heavy chain 7; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle beta isoform; myosin, heavy polypeptide 7, cardiac muscle, beta; Myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A
Gene Aliases: CMD1S; CMH1; MPD1; MYH7; MYHCB; SPMD; SPMM
UniProt ID: (Human) P12883
Entrez Gene ID: (Human) 4625
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.