Invitrogen
Manufacturer Code:MA527820
Catalog # PIMA527820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT565 |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human MYH6. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha cardiac MHC; alpha myosin; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; MyHC-alpha; Myosin heavy chain 6; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; Myosin-6; SSS3 ASD3 CMD1EE MYHCA MYHC myosin heavy chain 6 cardiac muscle alpha alpha-MHC CMH14
Gene Aliases: A830009F23Rik; AA517445; alpha-MHC; Myh6; Myhc-a; Myhca
UniProt ID: (Mouse) Q02566
Entrez Gene ID: (Mouse) 17888
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.