Novus Biologicals
Manufacturer Code:NB300284G
Catalog # NB300284G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 17593 |
Immunogen | Full length native protein (purified) (Human). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha-MHC ASD3 cardiomyopathy CMD1EE CMH14 Hypertrophic 1 MYHC MYHCA myHC-alpha Myosin Heavy Chain 6 Myosin Heavy Chain Cardiac Muscle Alpha Isoform Myosin Heavy Chain 6 Cardiac Muscle Alpha Myosin Heavy Polypeptide 6 Cardiac Muscle Alpha (Cardiomyopathy Hypertrophic 1) myosin-6 SSS3; MyHC-alpha; Myosin heavy chain 6; Myosin heavy chain, cardiac muscle alpha isoform; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); Myosin-6
Gene Aliases: alpha-MHC; ASD3; CMD1EE; CMH14; MYH6; MYHC; MYHCA; SSS3
UniProt ID: (Human) P13533
Entrez Gene ID: (Human) 4624
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.