Invitrogen
Manufacturer Code:PA597224
Catalog # PIPA597224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MYH6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha cardiac MHC; alpha myosin; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; MyHC-alpha; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); Myosin-6
Gene Aliases: A830009F23Rik; AA517445; alpha-MHC; ASD3; CMD1EE; CMH14; MYH6; MYHC; Myhc-a; MYHCA; SSS3
UniProt ID: (Human) P13533, (Mouse) Q02566
Entrez Gene ID: (Human) 4624, (Mouse) 17888, (Rat) 29556
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.