Abnova
Manufacturer Code:PAB20052
Catalog # 89-121-553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MYH6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MYH6.
For Research Use Only
Protein Aliases: alpha-MHC; ASD3; MYHC; MyHC-alpha; MYHCA; Myosin heavy chain 6; Myosin heavy chain, cardiac muscle alpha isoform; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); Myosin-6
Gene Aliases: alpha-MHC; ASD3; CMD1EE; CMH14; MYH6; MYHC; MYHCA; SSS3
UniProt ID: (Human) P13533
Entrez Gene ID: (Human) 4624
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.