Invitrogen
Manufacturer Code:PA567951
Catalog # PIPA567951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthesized peptide derived from human Myosin Skeletal Muscle. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2A myosin heavy chain; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MyHC-2a; MyHC-IIa; MYHSA1 MYHa MyHC-2X/D MyHC-2x myHC-IIx/d myosin heavy chain 1 myosin heavy chain 2x myosin heavy chain IIx/d myosin heavy chain skeletal muscle adult 1 myosin heavy polypeptide 1 skeletal muscle adult myosin-1; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain IIa; myosin heavy chain type IIx; Myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; Myosin-2; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain
Gene Aliases: IBM3; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIA; Myhs-f; Myhs-f1; MYHSA2; Myhsf1; MYPOP
UniProt ID: (Human) Q9UKX2
Entrez Gene ID: (Human) 4620, (Rat) 691644, (Mouse) 17882
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.