Proteintech
Manufacturer Code:207161AP150UL
Catalog # 50-173-3049
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MHC16 MYH14 myosin Myosin 14 Myosin heavy chain 14 myosin heavy chain 14 NMHC II C non-muscle II c; MYH14 variant protein; Myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy chain 14; myosin, heavy polypeptide 14; Myosin-14; NMHC II-C; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain; nonmuscle myosin heavy chain II-C
Gene Aliases: 2400004E04Rik; DFNA4; DFNA4A; FP17425; II-C; KIAA2034; MHC16; MYH14; MYH17; myosin; NHMCII; NMHC II-C; NMHC-II-C; PNMHH
UniProt ID: (Human) Q7Z406, (Mouse) Q6URW6
Entrez Gene ID: (Human) 79784, (Mouse) 71960, (Rat) 308572
Molecular Function:
G-protein modulator
actin binding motor protein
actin family cytoskeletal protein
cell junction protein
cytoskeletal protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.