Novus Biologicals
Manufacturer Code:NBP187743
Catalog # NBP187743
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:PDVEECAPNTGQCHLCLPPSEPWDQTLGVVNFPRKASRKPPREESSATCVLEQPGALGAQILLVQRPNSGLLAGLWEFPSVTWEPSEQLQRKALLQELQRWA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A/G-specific adenine DNA glycosylase; Adenine DNA glycosylase; CYP2C EC 3.2.2 EC 3.2.2.- hMYH MGC4416 mutY (E. coli) homolog MutY homolog mutY homolog (E. coli) Muyth MYHA/G-specific adenine DNA glycosylase; hMYH; MutY homolog; mutY-like protein
Gene Aliases: MUTYH; MYH
UniProt ID: (Human) Q9UIF7
Entrez Gene ID: (Human) 4595
Molecular Function:
DNA binding protein
DNA glycosylase
endodeoxyribonuclease
glycosidase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.