Novus Biologicals
Manufacturer Code:NBP272855JF549
Catalog # N272855J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G5 |
Immunogen | Full length recombinant protein of human MYF-5 (NP_005584) produced in E.coli. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: bHLHc2; BHLHC2 bHLHc2Class C basic helix-loop-helix protein 2 myf-5 myogenic factor 5; Class C basic helix-loop-helix protein 2; Myf-5; Myogenic factor 5
Gene Aliases: BHLHC2; MYF5
UniProt ID: (Human) P13349
Entrez Gene ID: (Human) 4617
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.