R&D Systems
Manufacturer Code:IC4027V100UG
Catalog # IC4027V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 593128 |
Immunogen | <EM>E. coli</EM>-derived recombinant human MYF-5 Met1-Leu255 Accession # P13349 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: bHLHc2; BHLHC2 bHLHc2Class C basic helix-loop-helix protein 2 myf-5 MYF5 myogenic factor 5; Class C basic helix-loop-helix protein 2; Myf-5; Myogenic factor 5
Gene Aliases: BHLHC2; MYF5
UniProt ID: (Human) P13349
Entrez Gene ID: (Human) 4617
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.