Proteintech
Manufacturer Code:111511AP150UL
Catalog # 50-172-6919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MYEOV Fusion Protein Ag1498 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: myeloma overexpressed (in a subset of t(11;14) positive multiple myelomas); myeloma overexpressed gene (in a subset of t(11;14) positive multiple myelomas); Myeloma-overexpressed gene protein; MYEOV OCIM Oncogene in multiple myeloma; Oncogene in multiple myeloma
Gene Aliases: MYEOV; OCIM
UniProt ID: (Human) Q96EZ4
Entrez Gene ID: (Human) 26579
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.