Invitrogen
Manufacturer Code:PA599796
Catalog # PIPA599796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MYCBP corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5730488M09Rik AMY1 Amy-1 Associate of Myc 1 associate of myc-1 AW552132 AW742590 c-myc binding protein C-Myc-binding protein Myc binding protein Mycbp; AMY-1; Associate of Myc 1; associate of myc-1; c-myc binding protein; C-Myc-binding protein
Gene Aliases: 5730488M09Rik; AMY-1; AMY1; AW552132; AW742590; MYCBP
UniProt ID: (Human) Q99417, (Mouse) Q9EQS3
Entrez Gene ID: (Human) 26292, (Mouse) 56309
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.