Invitrogen
Manufacturer Code:PA540439
Catalog # PIPA540439
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MYCBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMY-1; Associate of Myc 1; associate of myc-1; c-myc binding protein; C-Myc-binding protein
Gene Aliases: 5730488M09Rik; AMY-1; AMY1; AW552132; AW742590; MYCBP
UniProt ID: (Human) Q99417, (Bovine) Q2TBP7, (Mouse) Q9EQS3
Entrez Gene ID: (Dog) 475323, (Horse) 100068904, (Human) 26292, (Bovine) 539291, (Rabbit) 100343476, (Guinea pig) 100715652, (Mouse) 56309, (Rat) 100361133
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.