Novus Biologicals
Manufacturer Code:NBP189101
Catalog # NBP189101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LVALYEEPEKPNSALDFLKHHLGAATPENPEIELLRLELAEMKEKYEAIVEE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMY-1; AMY-1AMY1 Associate of Myc 1 associate of myc-1 c-myc binding protein C-Myc-binding protein FLJ41056; Associate of Myc 1; associate of myc-1; c-myc binding protein; c-Myc-binding protein
Gene Aliases: AMY-1; AMY1; MYCBP
UniProt ID: (Human) Q99417
Entrez Gene ID: (Human) 26292
Molecular Function: transcription cofactor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.