Abnova
Manufacturer Code:MAB7910
Catalog # 89-113-230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | LF10E3 |
Immunogen | A synthetic peptide corresponding to human MYC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of MYC.
For Research Use Only
Protein Aliases: avian myelocytomatosis viral oncogene homolog; bHLHe39; c-Myc; Class E basic helix-loop-helix protein 39; Myc proto-oncogene protein; myc-related translation/localization regulatory factor; Proto-oncogene c-Myc; Transcription factor p64; v-myc myelocytomatosis viral oncogene homolog
Gene Aliases: BHLHE39; c-Myc; MRTL; MYC; MYCC
UniProt ID: (Human) P01106
Entrez Gene ID: (Human) 4609
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.