Invitrogen
Manufacturer Code:PA571701
Catalog # PIPA571701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 189-218 amino acids from the N-terminal region of human MYBPC3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-protein, cardiac muscle isoform; cardiac C-protein; Cardiac MyBP-C; CMH4 C-protein cardiac muscle isoform FHC MYBP-C Cardiac MyBP-C CMH4 C-protein cardiac muscle isoform DKFZp779E1762 FHC MYBP-C myosin binding protein C cardiac myosin-binding protein C cardiac myosin-binding protein C cardiac-type; Myosin-binding protein C, cardiac-type; truncated cardiac myosin-binding protein C
Gene Aliases: CMD1MM; CMH4; FHC; LVNC10; MYBP-C; MYBPC3
UniProt ID: (Human) Q14896, (Rat) P56741
Entrez Gene ID: (Human) 4607, (Rat) 295929, (Mouse) 17868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.